missing translation for 'onlineSavingsMsg'
Learn More

CTRP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14023-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892484

  • 1656.73 SEK / 20µL
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of mouse CTRP4 have been reported.
Specifications

Specifications

CTRP4
Polyclonal
Unconjugated
C1QTNF4
C1QTNF4, CTRP4, ZACRP4
Rabbit
Antigen Affinity Chromatography
RUO
114900, 311184, 67445
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R066, Q9BXJ3
C1qtnf4
CTRP4 Fusion Protein Ag5147
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers