missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CTSE (Human) Recombinant Protein
Human CTSE full-length ORF ( AAH42537, 18 a.a. - 396 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001510-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: QGSLHRVPLRRHPTLKKKLRARSQLSEFWKSHNLDMIQFTESCSMDQSAKEPLINYLDMEYFGTISIGSPPQNFTVIFDTGSSNLWVPSVYCTSPACKTHSRFQPSQSSTYSQPGQSFSIQYGTGSLSGIIGADQVSVEGLTVVGQQFGESVTEPGQTLVDAEFDGILGLGYPSLAVGGVTPVFDNMMAQNLVDLPMFSVYMSSNPEGGAGSELIFGGYDHSHFSGSLNWVPVTKQAYWQIALDNIQVGGTVMFCSEGCQAIVDTGTSLITGPSDKIKQLQNAIGAAPVDGEYAVECANLNVMPDVTFTINGVPYTLSPTAYTLLDFVDGMQFCSSGFQGLDIHPPAGPLWILGDVFIRQFYSVFDRGNNRVGLAPAVP
Specifications
AAH42537 | |
cathepsin E | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTSE | |
GST |
1510 | |
Wheat germ expression system | |
10 μg | |
CATE | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |