missing translation for 'onlineSavingsMsg'
Learn More

CUL4A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10693-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827243

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
Specifications

Specifications

CUL4A
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13619
CUL 4A, CUL4A, cullin 4A, Cullin-4A
Rabbit
IgG
150 μL
-20°C
Primary
8451
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CUL4A
Liquid
CUL4A
CUL4A Fusion Protein Ag1030
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers