missing translation for 'onlineSavingsMsg'
Learn More

Cupferron, Thermo Scientific Chemicals

Catalog Number p-94502
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
135-20-6
C6H9N3O2
155.16
MFCD00078422
NJVHCUNZAMFQNA-UHFFFAOYSA-N
cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline
2724103
azanium;N-oxido-N-phenylnitrous amide
N.ON(N=O)C1=CC=CC=C1
This item is not returnable. View return policy
135-20-6
C6H9N3O2
155.16
MFCD00078422
NJVHCUNZAMFQNA-UHFFFAOYSA-N
cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline
2724103
azanium;N-oxido-N-phenylnitrous amide
N.ON(N=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150.0°C to 155.0°C
Ignition Residue 0.05% max
Linear Formula C6H4N(ONH4)NO
Quantity 25 g
Beilstein 16,I,395
Merck Index 15,2612
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether
Formula Weight 155.16
Grade ACS Reagent
Chemical Name or Material Cupferron
compliance-icons compliance-icons
Product Identifier
  • Cupferron
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'