missing translation for 'onlineSavingsMsg'
Learn More

Cuprolinic Blue (>90%), TRC

Product Code. 30367150
Change view
Click to view available options
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30367150 2.5 mg
30366090 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30367150 Supplier Toronto Research Chemicals Supplier No. TRCC8348002.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41276-95-3
Molekylformel C32 H26 N12 . 4 C H3 O4 S . Cu
Molekylvikt (g/mol) 1086.56
Synonym (SP-4-1)-[1,8,15,22-tetramethyl-29H,31H-tetrapyrido[2,3-b:2',3'-g:2'',3''-l:2''',3'''-q]porphyraziniumato(2-)-κN29,κN30,κN31,κN32]copper(4+) Methyl Sulfate (1:4), [1,8,15,22-tetramethyltetrapyrido[2,3-b:2',3'-g:2'',3''-l:2''',3'''-q]porphyraziniumato(2-)]copper(4+) Ion Tetrakis(methyl sulfate), 29H,31H-Tetrapyrido[2,3-b:2',3'-g:2'',3''-l:2''',3'''-q]porphyrazine, copper(4+) Deriv., Cuproline Blue, Quinolinic phthalocyanine
IUPAC-namn copper;methyl sulfate;5,14,23,32-tetramethyl-2,11,20,29,37,39-hexaza-5,14,23,32-tetrazonia-38,40-diazanidanonacyclo[28.6.1.13,10.112,19.121,28.04,9.013,18.022,27.031,36]tetraconta-1,3,5,7,9,11,13(18),14,16,19(39),20,22(27),23,25,28,30(37),31(36),32,34-nonadecaene
LEDER [Cu].COS(=O)(=O)[O-].COS(=O)(=O)[O-].COS(=O)(=O)[O-].COS(=O)(=O)[O-].C[n+]1cccc2c3nc4nc(nc5[nH]c(nc6nc(nc([nH]3)c12)c7ccc[n+](C)c67)c8ccc[n+](C)c58)c9ccc[n+](C)c49

Specifications

Kemiskt namn eller material Cuprolinic Blue
Kvantitet 2.5 mg
InChI formel InChI=1S/C32H25N12.4CH4O4S.Cu/c1-41-13-5-9-17-21(41)29-33-25(17)38-30-23-19(11-7-15-43(23)3)27(35-30)40-32-24-20(12-8-16-44(24)4)28(36-32)39-31-22-18(26(34-31)37-29)10-6-14-42(22)2;4*1-5-6(2,3)4;/h5-16H,1-4H3,(H,33,34,35,36,37,38,39,40);4*1H3,(H,2,3,4);/q+3;;;;;/p-3
Formel vikt 1085.0708
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.