Learn More
Macherey-Nagel™ Cuprotesmo Qualitative Test Paper
Brand: Macherey-Nagel™ 90601
Additional Details : Weight : 0.01000kg
Specifications
Yellow/White to Pink/Purple | |
For detection of Copper, Copper ions | |
Qualitative Test Paper |
Sheet | |
40 Pack |