Learn More
Macherey-Nagel™ Cuprotesmo Qualitative Test Paper
Brand:  Macherey-Nagel™ 90601
Additional Details : Weight : 0.01000kg
Product Code. 11316504
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Yellow/White to Pink/Purple
For detection of Copper, Copper ions
Qualitative Test Paper
Sheet
40 Pack