missing translation for 'onlineSavingsMsg'
Läs mer

Curcumin, Sigma-Aldrich™

Produktkod. 32238465
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
32238465 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 32238465 Leverantör Sigma Aldrich Leverantörsnummer 8203540002
 Begränsad artikel
Beräknad leverans: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Curcumin
CAS 458-37-7
Smältområde 170°C to 175°C
Molekylformel [4-(OH)-3-(CH3O)C6H3CH=CHCO]2CH2
Kvantitet 2 g
Synonym Curcumin, 1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Turmeric yellow, Diferuloylmethane, Curcumin
Molekylvikt (g/mol) 368.38
Kvalitet Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.