missing translation for 'onlineSavingsMsg'
Läs mer

Curvulin, TRC

Produktkod. 30180158
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180158 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30180158 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC8445051MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Curvulin
CAS 19054-27-4
Molekylformel C12H14O5
Kvantitet 1 mg
InChI formel InChI=1S/C12H14O5/c1-3-17-11(16)5-8-4-9(14)6-10(15)12(8)7(2)13/h4,6,14-15H,3,5H2,1-2H3
LEDER CCOC(=O)Cc1cc(O)cc(O)c1C(=O)C
IUPAC-namn ethyl 2-(2-acetyl-3,5-dihydroxyphenyl)acetate
Molekylvikt (g/mol) 238.24
Formel vikt 238.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.