Learn More
3M™ Safety-Walk™ Cushion Matting 5100
Rigid, vinyl Z web construction provides firm support and allows cart traffic for a multifunctional work area. Available in various sizes, with optional glued on edging.
Brand: 3M™ Safety-Walk™ 7100116734
Additional Details : Weight : 18.28430kg
Description
- Cushioned matting with an extended ridge design
- Offers excellent slip resistance in wet areas
- Vinyl composition provides excellent resistance to a variety of chemicals, greases, and detergents found in industrial or food service environments
- Open web construction to allow cart traffic and good water drainage
- Anti-microbial coating protects against mildew, fungus, and odors
Specifications
10 ft. | |
3 ft. | |
1 Ea. | |
3.1 m |
Black | |
Cushion Mat | |
0.914 m |