Läs mer
Custom Primer
Produktkod. 10860211
Produktkod. 10860211 Leverantör Invitrogen Leverantörsnummer 11732039
 Begränsad artikel
Beräknad leverans: 02-11-2026
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).