Läs mer
Custom Primer
Produktkod. 10764264
Produktkod. 10764264 Leverantör Invitrogen Leverantörsnummer 11745023
 Begränsad artikel
Beräknad leverans: 09-11-2026
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).