Learn More
Ambion™ Customer-Defined Pre-miR™ miRNA Precursor
Customer-Defined Pre-miR™ miRNA Precursor
Brand:  Ambion™ AM17103
Additional Details : Weight : 0.01000kg
Product Code. 15521531
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Pre-miR™ miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs. Use our customization tool to design the ideal mimic for your miRNA sequence, or select from among predesigned products.
Specifications
Specifications
Room temperature
Pre-miRNA Precursor, 20nmol, Small, Chemically Modified Double-Stranded RNA Molecules Designed to Mimic Endogenous Mature miRNAs, Use Our Customization Tool to Design the Ideal Mimic for Your miRNA Sequence
Pre-miRNA Precursor
20 nmol