missing translation for 'onlineSavingsMsg'
Learn More

CUTC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15030-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824004

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the CUT family of copper transporters are associated with copper homeostasis and are involved in the uptake, storage, delivery, and efflux of copper (Gupta et al., 1995 ; Li et al., 2005 ).
Specifications

Specifications

CUTC
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D8X1, Q9NTM9
CGI 32, CUTC, RP11 483F11.3
Rabbit
IgG
20 μL
-20°C
Primary
361760, 51076, 66388
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
CUTC
Liquid
CUTC
CUTC Fusion Protein Ag2126
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers