Läs mer
Ansell Cutstar Medium Duty Cut Protection Gloves
Produktkod. 16865280 Handla allt Ansell Produkter
Storlek:
7
8
9
10
Förpackningsstorlek:
12 par
Produktkod. 16865280
Brand: Ansell B047070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Lined with high performance fibers using Techcor™ Knitting Technology for increased cut protection. Plastoprene dots on the palm and fingers enhance grip capabilities.
  • Liner constructed with high performance fibers, using Techcor™ Knitting Technology
  • Plastoprene dots on palm and fingers enhance grip capabilities
  • Protection against cuts and abrasions
TRUSTED_SUSTAINABILITY
Specifikationer
ANSI Abrasion Rating ANSI 4
Certifieringar/efterlevnad (Europa) EN 420:2003 + A1:2009, REACH Compliant
Beläggningsmaterial PVC
Färg White
Betyg för skärmotstånd ANSI A3
Hand Specific
Material Synthetic Fiber
Storlek 7
EN388 Standard 3X42B
För användning med (applikation) Assembly
Handske manschettstil Knit
Serie Cutstar