missing translation for 'onlineSavingsMsg'
Learn More

Ansell Cutstar Medium Duty Cut Protection Gloves

Catalog Number 16895280
Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Lined with high performance fibers using Techcor™ Knitting Technology for increased cut protection. Plastoprene dots on the palm and fingers enhance grip capabilities.

  • Liner constructed with high performance fibers, using Techcor™ Knitting Technology
  • Plastoprene dots on palm and fingers enhance grip capabilities
  • Protection against cuts and abrasions
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material PVC
Color White
Cut Resistance Rating ANSI A3
Hand Specific
Material Synthetic Fiber
Size 10
EN388 Standard 3X42B
For Use With (Application) Assembly
Glove Cuff Style Knit
Series Cutstar
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'