missing translation for 'onlineSavingsMsg'
Learn More

Ansell Cutstar Medium Duty Cut Protection Gloves

Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Lined with high performance fibers using Techcor™ Knitting Technology for increased cut protection. Plastoprene dots on the palm and fingers enhance grip capabilities.

  • Liner constructed with high performance fibers, using Techcor™ Knitting Technology
  • Plastoprene dots on palm and fingers enhance grip capabilities
  • Protection against cuts and abrasions
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material PVC
Color White
Cut Resistance Rating ANSI A3
Hand Specific
Material Synthetic Fiber
Size 9
EN388 Standard 3X42B
For Use With (Application) Assembly
Glove Cuff Style Knit
Series Cutstar
Show More Show Less