Learn More
WTW™ Cuvette Cells for PhotoLab™ Photometers
WTW™ Cells for PhotoLab™ Photometers are ideal for routine water and other analytics in the UV-VIS and VIS range.
1895.00 SEK - 3679.08 SEK
Specifications
Type | Cuvette |
---|---|
Disposable | No |
Material | Quartz |
For Use With (Equipment) | Photometers |
Shape | Rectangle |
Product Code | Brand | Path Length | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Path Length | Price | Quantity & Availability | |||||
16420721
|
WTW™
250606 |
10 mm |
1895.00 SEK
Each |
Estimated Shipment: 03-09-2024
Log in to see stock. |
|||||
16430721
|
WTW™
250615 |
50 mm |
3679.08 SEK
Each |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
- Cells with matt side surface are suitable for the automatic cell detection of the photoLab™ filter and spectral photometer
- All glass and quartz cuvettes have placement marks to locate them in the photometer in the same orientation for even higher precision
- High quality optical glass for VIS measurements, high quality quartz glass for UV-VIS
Specifications
Cuvette | |
Quartz | |
Rectangle |
No | |
Photometers |