missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CUX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115066
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Antibody detects endogenous levels of total CUTL1.
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifikationer
| CUX1 | |
| Polyclonal | |
| Unconjugated | |
| CUX1 | |
| CASP; CCAAT displacement protein; CDP; CDP/Cut; CDP1; CDP2; Cdpl1; Clox; COY1; cut homolog; cut like homeobox 1; CUTL1; cut-like 1; cut-like homeobox 1; Cux; Cux/CDP; Cux/CDP homeoprotein; CUX1; Cux-1; golgi integral membrane protein 6; GOLIM6; Homeobox protein cut-like 1; homeobox protein cux-1; Kiaa4047; Nbla10317; p100; p110; p200; p75; Protein CASP; putative protein product of Nbla10317 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13047, 1523 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P39880, P70403, Q13948 | |
| CUX1 | |
| A synthesized peptide derived from human CUX1(Accession P39880), corresponding to amino acid residues L1189-Q1239. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido