Learn More
IKA CV Package for ElectraSyn 2.0
Cyclic voltammetry electrode package suitable for use with the ElectraSyn 2.0.
Brand:  IKA 0040002864
Additional Details : Weight : 0.05000kg
Product Code. 15810965
  • 8400.00 SEK / Each
Estimated Shipment: 29-07-2022
to see stock.
Add to basket
Description
Description
  • Designed for use with the ElectraSyn 2.0 and ElectraSyn 2.0 Pro.
  • Includes: CV Glassy Carbon, CV Platinum, Reference Electrode.
Specifications
Specifications
Electrochemistry Kit