Learn More
Tocris Bioscience™ CW 008
GREENER_CHOICE
PKA signaling activator; promotes osteogenesis from hMSCs
Brand:  Tocris Bioscience™ 5495/50
Encompass
Product Code. 15776886
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
CW 008 is a cAMP/PKA/CREB pathway activator. Promotes osteogenic differentiation of human bone marrow-derived MSCs in vitro. Increases bone mass and bone volume density in ovariectomized mice.
Specifications
Specifications
4-Fluoro-N-[5-fluoro-6-(5-methoxypyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-b]pyridin-3-yl]benzamide
1134613-19-6
50 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.