Learn More
Tocris Bioscience™ CW 008
PKA signaling activator; promotes osteogenesis from hMSCs
Brand: Tocris Bioscience™ 5495/50
This item is not returnable.
View return policy
Description
CW 008 is a cAMP/PKA/CREB pathway activator. Promotes osteogenic differentiation of human bone marrow-derived MSCs in vitro. Increases bone mass and bone volume density in ovariectomized mice.Specifications
| 4-Fluoro-N-[5-fluoro-6-(5-methoxypyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-b]pyridin-3-yl]benzamide | |
| 1134613-19-6 | |
| 50 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.