missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX3CL1 Mouse anti-Human, Clone: 2F5D5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60339-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the CX3C subgroup of chemokines, characterized by the number of amino acids located between the conserved cysteine residues. This is the only member of the CX3C subgroup, which contains three amino acids between cysteine residues, resulting in a Cys-X-X-X-Cys configuration. The encoded protein contains an extended mucin-like stalk with a chemokine domain on top, and exists in both a membrane-anchored form where it acts as a binding molecule, or, in soluble form, as a chemotactic cytokine. The mature form of this protein can be cleaved at the cell surface, yielding different soluble forms that can interact with the G-protein coupled receptor, C-X3-C motif chemokine receptor 1 gene product. This gene plays a role in a wide range of diseases, including cancer, vasculitis, neuropathies, atherosclerosis, inflammatory diseases, and in human immunodeficiency virus infections.Specifications
CX3CL1 | |
Monoclonal | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78423 | |
CX3CL1 | |
CX3CL1 Fusion Protein Ag0161 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2F5D5 | |
Unconjugated | |
CX3CL1 | |
ABCD 3, C X3 C motif chemokine 1, C3Xkine, CX3CL1, CXC3, CXC3C, FKN, fractalkine, neurotactin, NTN, NTT, SCYD1, Small inducible cytokine D1 | |
Mouse | |
Protein G | |
RUO | |
6376 | |
-20°C | |
Liquid |