missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CXCL1 (Human) Recombinant Protein (P01)

Human CXCL1 full-length ORF with GST-tag at N-terminal

Brand:  Abnova™ H00002919-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135791

  • 6845.00 SEK / 25µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Human CXCL1 full-length ORF ( AAH11976, 1 a.a. - 107 a.a.) recombinant protein with GST-tag at N-terminal

  • Theoretical MW: 37.51kDa
  • Preparation Method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Best use within three months from the date of receipt of this protein

Enzyme Linked Immunosorbent Assay, Western Blotting, Antibody Production, Protein Array

Specifications

Specifications

AAH11976
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
37.51
8
Glutathione Sepharose 4 Fast Flow
25 ug
MARAALSAAPSNPRLLRVALLLLLLVAAGRRAAGASVATELRCQCLQTLQGIHPKNIQSVNVKSPGPHCAQTEVIATLKNGRKACLNPASPIVKKIIEKMLNSDKSN
FSP/GRO1/GROa/MGSA/MGSA-a/NAP-3/SCYB1
CXCL1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
2919
CXCL1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CXCL1
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers