Läs mer
PeproTech CXCL1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
2250.00 SEK - 37440.00 SEK
Specifikationer
| Antigen | CXCL1 |
|---|---|
| Koncentration | 0.1-1.0 mg/mL |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Biotin |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
17884153
|
PeproTech
500-P115BT-25UG |
25 μg |
2250.00 SEK
25µg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
17883843
|
PeproTech
500-P115BT-50UG |
50 μg |
2890.00 SEK
50µg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
17804373
|
Invitrogen
500-P115BT-1MG |
2 x 500 μg | |||||||
|
30054734
|
PeproTech
500-P115BT-500UG |
500 μg |
19980.00 SEK
500µg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
AA Sequence of recombinant protein: APIANELRCQ CLQTMAGIHL KNIQSLKVLP SGPHCTQTEV IATLKNGREA CLDPEAPLVQ KIVQKMLKGV PK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine KC (CXCL1). Anti-Murine KC (CXCL1)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine KC (CXCL1) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine KC (CXCL1) (500-P115) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine KC (CXCL1). Western Blot: To detect mKC by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mKC is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P115BT-1MG will be provided as 2 x 500 μg
CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.Specifikationer
| CXCL1 | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Mouse | |
| P12850 | |
| 14825 | |
| E.coli-derived Recombinant Murine KC (CXCL1). | |
| Antigen affinity chromatography | |
| Cxcl1 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| alpha-chemokine; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); Cinc1; CINC-1; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine ligand 1; CXCL; CXCL1; CXCL-1; Cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; Fsp; Gro; GRO a; GRO α; Gro1; GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); Growth-regulated alpha protein; GROα; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC/GR)-alpha; KC/GRO-alpha; KC-T; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; MGSA-a; N51; NAP3; NAP-3; Neutrophil-activating protein 3; platelet-derived growth factor-inducible protein KC; SCYB1; secretory protein N51 | |
| Cxcl1 | |
| Primary | |
| -20°C |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.