missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ CXCL10 (IP-10) Monoclonal Antibody (4NY8UN), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Produktkod. 16696414
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16696414 25 Tests
16606424 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16696414 Leverantör Invitrogen™ Leverantörsnummer 53974441

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen CXCL10 (IP-10)
Användningsområden Flow Cytometry
Klassificering Monoclonal
Klona 4NY8UN
Koncentration 5 μL/Test
Konjugera Alexa Fluor 488
Formulering PBS with BSA and 0.09% sodium azide; pH 7.2
Gen CXCL10
Genaccessionsnr. P02778
Gene Alias 10 kDa interferon gamma-induced protein; 10 kDa interferon-gamma induced protein precursor; C Cmotif chemokine; C X C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; CRG-2; CXC; CXC chemokine; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; CXCL10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; gamma-IP10; gIP-10; H-IP-10; IFI10; Inp10; interferon activated gene 10; interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; protein Mob-1; Scyb10; similar to Small inducible cytokine B10 precursor (CXCL10) (10 kDa interferon-gamma induced protein) (Gamma-IP10) (IP-10); small inducible cytokine B subfamily (Cys-X-Cys) member 10; small inducible cytokine B subfamily (Cys-X-Cys), member 10; small inducible cytokine subfamily B (Cys-X-Cys), member 10; small-inducible cytokine B10
Gensymboler CXCL10
Värd art Mouse
Reningsmetod Affinity chromatography
Kvantitet 25 Tests
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 3627
Målarter Human
Innehåll och lagring 4°C, store in dark, DO NOT FREEZE!
Produkttyp Antibody
Form Liquid
Isotyp IgG2b κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.