missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL12/SDF-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CXCL12/SDF-1 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827294
|
Proteintech
17402-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817294
|
Proteintech
17402-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody can detect all the isoforms of CXCL12/SDF1.
CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
CXCL12/SDF-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P48061 | |
6387 | |
CXCL12/SDF-1 Fusion Protein Ag11379 | |
Primary | |
-20°C | |
CXCL12 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C X C motif chemokine 12, CXCL12, hIRH, hSDF 1, IRH, PBSF, SCYB12, SDF 1, SDF 1a, SDF 1b, SDF1, SDF1A, SDF1B, Stromal cell derived factor 1, TLSF a, TLSF b, TPAR1 | |
CXCL12 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |