missing translation for 'onlineSavingsMsg'
Learn More

CXCL13/BCA1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10927-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827643

  • 1236.20 SEK / 20µL
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Chemotactic for B-lymphocytes but not for T-lymphocytes. monocytes and neutrophils. Does not induce calcium release in B-lymphocytes. Binds to BLR1/CXCR5. B lymphocyte chemoattractant. independently cloned and named Angie.
Specifications

Specifications

CXCL13/BCA1
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43927
ANGIE, ANGIE2, B cell attracting chemokine 1, B lymphocyte chemoattractant, BLC, BLR1L, C X C motif chemokine 13, CXC chemokine BLC, CXCL13, SCYB13, Small inducible cytokine B13
Rabbit
IgG
20 μL
-20°C
Primary
10563
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
CXCL13
Liquid
CXCL13
CXCL13/BCA1 Fusion Protein Ag1358
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers