missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL16 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | CXCL16 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16832324
|
Proteintech
13902-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16822324
|
Proteintech
13902-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CXCL16 acts as a scavenger receptor on macrophages, which specifically binds to OxLDL (oxidized low density lipoprotein), suggesting that it may be involved in pathophysiology such as atherogenesis. Induces a strong chemotactic response. Induces calcium mobilization. Binds to CXCR6/Bonzo.Specifications
CXCL16 | |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q9H2A7 | |
58191 | |
CXCL16 Fusion Protein Ag4872 | |
Primary | |
-20°C | |
CXCL16 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C X C motif chemokine 16, CXCL16, CXCLG16, SCYB16, Small inducible cytokine B16, SR PSOX, SRPSOX, Transmembrane chemokine CXCL16 | |
CXCL16 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |