missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CXCL9 (Human) Recombinant Protein

Human CXCL9 full-length ORF ( AAH63122, 23 a.a. - 125 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004283-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137831

  • 4515.00 SEK / 10µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: TPVVRKGRCSCISTNQGTIHLQSLKDLKQFAPSPSCEKIEIIATLKNGVQTCLNPDSADVKELIKKWEKQVSQKKKQKNGKKHQKKKVLKVRKSQRSRQKKTT
Specifications

Specifications

AAH63122
chemokine (C-X-C motif) ligand 9
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CXCL9
GST
4283
Wheat germ expression system
10 μg
CMK, Humig, MIG, SCYB9, crg-10
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers