missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55450-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to CXCR1.
CXCR1 is a member of the G-protein-coupled receptor family. This protein is a receptor for interleukin 8 (IL8). It binds to IL8 with high affinity, and transduces the signal through a G-protein activated second messenger system. Knockout studies in mice suggested that this protein inhibits embryonic oligodendrocyte precursor migration in developing spinal cord. This gene, IL8RB, a gene encoding another high affinity IL8 receptor, as well as IL8RBP, a pseudogene of IL8RB, form a gene cluster in a region mapped to chromosome 2q33-q36.Specifications
CXCR1 | |
Polyclonal | |
Unconjugated | |
CXCR1 | |
C C, C C CKR 1, CD128, CD181, CDw128a, CKR 1, CMKAR1, CXC R1, CXCR 1, CXCR1, IL 8 receptor type 1, IL 8R A, IL8R1, IL8RA, IL8RBA, interleukin 8 receptor, alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227288, 3577 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P25024, Q810W6 | |
CXCR1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |