missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CXorf2 (Human) Recombinant Protein
Human CXorf2 full-length ORF ( AAH26183, 1 a.a. - 410 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH26183 |
---|---|
Gene ID (Entrez) | 1527 |
Name | testis expressed 28 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16123791
|
Abnova™
H00001527-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16133791
|
Abnova™
H00001527-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MVLKAEHTRSPSATLPSNVPSCRSLSSSEDGPSGPSSLADGGLAHNLQDSVRHRILYLSEQLRVEKASRDGNTVSYLKLVSKADRHQVPHIQQAFEKVNQRASATIAQIEHRLHQCHQQLQELEEGCRPEGLLLMAESDPANCEPPSEKALLSEPPEPGGEDGPVNLPHASRPFILESRFQSLQQGTCLETEDVAQQQNLLLQKVKAELEEAKRFHISLQESYHSLKERSLTDLQLLLESLQEEKCRQALMEEQVNGRLQGQLNEIYNLKHNLACSEERMAYLSYERAKEIWEITETFKSRISKLEMLQQVTQLEAAEHLQSRPPQMLFKFLSPRLSLATVLLVFVSTLCACPSSLISSRLCTCTMLMLIGLGVLAWQRWRAIPATDWQEWVPSRCRLYSKDSGPPADGP
Specifications
AAH26183 | |
testis expressed 28 | |
125% SDS-PAGE Stained with Coomassie Blue | |
CXorf2, fTEX | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
1527 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TEX28 | |
GST |