missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CXorf2 (Human) Recombinant Protein

Human CXorf2 full-length ORF ( AAH26183, 1 a.a. - 410 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00001527-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133791

  • 5030.41 SEK / 25µg

Estimated Shipment: 25-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MVLKAEHTRSPSATLPSNVPSCRSLSSSEDGPSGPSSLADGGLAHNLQDSVRHRILYLSEQLRVEKASRDGNTVSYLKLVSKADRHQVPHIQQAFEKVNQRASATIAQIEHRLHQCHQQLQELEEGCRPEGLLLMAESDPANCEPPSEKALLSEPPEPGGEDGPVNLPHASRPFILESRFQSLQQGTCLETEDVAQQQNLLLQKVKAELEEAKRFHISLQESYHSLKERSLTDLQLLLESLQEEKCRQALMEEQVNGRLQGQLNEIYNLKHNLACSEERMAYLSYERAKEIWEITETFKSRISKLEMLQQVTQLEAAEHLQSRPPQMLFKFLSPRLSLATVLLVFVSTLCACPSSLISSRLCTCTMLMLIGLGVLAWQRWRAIPATDWQEWVPSRCRLYSKDSGPPADGP
Specifications

Specifications

AAH26183
testis expressed 28
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TEX28
GST
1527
Wheat germ expression system
25 μg
CXorf2, fTEX
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates