Learn More
CXorf58 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556776
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FPKSKVTDIM DVVTMQDYVQ YRSFFDEAPA FSGGRNNSWR KLNLENIPRT MLMYDIVHYS ESGVISNRLR NEMKF Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 26%.
CXorf58 Polyclonal antibody specifically detects CXorf58 in Human samples. It is validated for Immunohistochemistry.Specifications
CXorf58 | |
Polyclonal | |
Unconjugated | |
CXorf58 | |
chromosome X open reading frame 58; CXorf58; Putative uncharacterized protein CXorf58 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254158 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LI9 | |
CXorf58 | |
Recombinant protein corresponding to Human CXorf58 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only