missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXXC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566138
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: ASLLANGHDLA AAMAVDKSNP TSKHKSGAVA SLLSKAERAT ELAAEGQLTL QQFAQSTEML KRVVQEHLPL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
CXXC5 Polyclonal antibody specifically detects CXXC5 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
CXXC5 | |
Polyclonal | |
Unconjugated | |
CXXC5 | |
4930415K17Rik; CF5; CXXC finger 5; CXXC finger 5 protein; CXXC finger protein 5; Cxxc5; CXXC-type zinc finger protein 5; HSPC195; Putative MAPK-activating protein PM08; putative NF-kappa-B-activating protein 102; Retinoid-inducible nuclear factor; RINF; TCCCIA00297; WID; WT1-induced Inhibitor of Dishevelled | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51523 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7LFL8 | |
CXXC5 | |
Recombinant Human CXXC5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only