Läs mer
Cyanamide-15N2, TRC
Produktkod. 30355429
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355429 1 mg
30353987 10 mg
30351661 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30355429 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9525031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21420-37-1
Molekylformel C H2 15N2
Molekylvikt (g/mol) 44.027
IUPAC-namn (15N)azanylformonitrile
LEDER [15NH2]C#[15N]
Specifikationer
Kemiskt namn eller material Cyanamide-15N2
Kvantitet 1 mg
InChI formel InChI=1S/CH2N2/c2-1-3/h2H2/i2+1,3+1
Formel vikt 44.016