Läs mer
Cyanazine Acid, TRC
Produktkod. 30138181
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138181 50 mg
30133346 100 mg
17950725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138181 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC95355050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36576-43-9
Molekylformel C9 H14 Cl N5 O2
Molekylvikt (g/mol) 259.69
Synonym N-[4-Chloro-6-(ethylamino)-1,3,5-triazin-2-yl]-2-methylalanine, 2-Chloro-4-(1-carboxy-1-methylethylamino)-6-(ethylamino)-1,3,5-triazine, Cyanazine III, Cyanazine acid, N-[4-chloro-6-(ethylamino)-1,3,5-triazin-2-yl]-2-methyl-Alanine;
IUPAC-namn 2-[[4-chloro-6-(ethylamino)-1,3,5-triazin-2-yl]amino]-2-methylpropanoic acid
LEDER CCNc1nc(Cl)nc(NC(C)(C)C(=O)O)n1
Specifikationer
Kemiskt namn eller material Cyanazine acid
Kvantitet 50 mg
InChI formel InChI=1S/C9H14ClN5O2/c1-4-11-7-12-6(10)13-8(14-7)15-9(2,3)5(16)17/h4H2,1-3H3,(H,16,17)(H2,11,12,13,14,15)
Formel vikt 259.0836