Learn More
SPEX CertiPrep™ Cyanide and Phenol QC Sample
Brand: SPEX CertiPrep™ 502
Additional Details : Weight : 0.09000kg
Specifications
Vial | |
Ambient | |
Yields up to 2 L after dilution |
Cyanide and Phenol Wastewater QC Sample | |
15 mL |