Läs mer
Cyanidin 3-Xyloside, TRC
Produktkod. 30136388
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136388 2.5 mg 2.5mg
30133179 5 mg -
30012735 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136388 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9537002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29761-24-8
Molekylformel C20 H19 O10 . Cl
Molekylvikt (g/mol) 454.81
Synonym 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-(D-xylopyranosyl)-1-benzopyrylium chloride (1:1)
IUPAC-namn (3R,4S,5R)-2-[2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-chromenylium-3-yl]oxytetrahydropyran-3,4,5-triol;chloride
LEDER [Cl-].O[C@@H]1COC(Oc2cc3c(O)cc(O)cc3[o+]c2c4ccc(O)c(O)c4)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material Cyanidin-3-xyloside chloride
Kvantitet 2.5 mg
InChI formel InChI=1S/C20H18O10.ClH/c21-9-4-12(23)10-6-16(30-20-18(27)17(26)14(25)7-28-20)19(29-15(10)5-9)8-1-2-11(22)13(24)3-8;/h1-6,14,17-18,20,25-27H,7H2,(H3-,21,22,23,24);1H/t14-,17+,18-,20?;/m1./s1
Formel vikt 454.0667