Läs mer
Cyanidin Chloride, TRC
Produktkod. 30136383
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136383 5 mg
17979444 1 mg
17977264 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136383 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9536455MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 528-58-5
Molekylformel C15 H11 O6 . Cl
Molekylvikt (g/mol) 322.7
Synonym 1-Benzopyrylium, 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-, chloride (1:1), 1-Benzopyrylium, 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-, chloride (9CI), Cyanidol (6CI), Flavylium, 3,3',4',5,7-pentahydroxy-, chloride (8CI), 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-1-benzopyrylium chloride, 3,3',4',5,7-Pentahydroxy-2-phenylbenzopyrylium chloride, 3,3',4',5,7-Pentahydroxyflavylium chloride, Cyanidin, Cyanidin chloride, Cyanidine, Cyanidol chloride, IdB 1027
IUPAC-namn 2-(3,4-dihydroxyphenyl)chromenylium-3,5,7-triol;chloride
LEDER [Cl-].Oc1cc(O)c2cc(O)c([o+]c2c1)c3ccc(O)c(O)c3
Specifikationer
Kemiskt namn eller material Cyanidin chloride
Kvantitet 5 mg
InChI formel InChI=1S/C15H10O6.ClH/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7;/h1-6H,(H4-,16,17,18,19,20);1H
Formel vikt 322.0244