Läs mer
Cyanidol 3-Glucoside, TRC
Produktkod. 30011344
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011344 5 mg
30004667 10 mg
30007049 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011344 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9877705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7084-24-4
Molekylformel C21 H21 O11 . Cl
Molekylvikt (g/mol) 484.84
Synonym 1-Benzopyrylium, 2-(3,4-dihydroxyphenyl)-3-(β-D-glucopyranosyloxy)-5,7-dihydroxy-, chloride (1:1), 1-Benzopyrylium, 2-(3,4-dihydroxyphenyl)-3-(β-D-glucopyranosyloxy)-5,7-dihydroxy-, chloride (9CI), Chrysanthemin (6CI,8CI), 3,3',4',5,7-Pentahydroxyflavylium chloride, 3-glucoside, 3-(β-D-Glucopyranosyloxy)-3',4',5,7-tetrahydroxyflavylium chloride, 3-O-(β-D-Glucopyranosyl)cyanidin, Asterin, Chrysanthenin, Chrysontemin, Cyanidin 3-O-glucoside, Cyanidin 3-O-glucoside chloride, Cyanidin 3-O-β-D-glucopyranoside, Cyanidin 3-O-β-glucopyranoside, Cyanidin 3-glucoside, Cyanidin 3-glucoside chloride, Cyanidin 3-monoglucoside, Cyanidin 3-β-D-glucopyranoside, Cyanidin 3-β-O-glucoside, Cyanidin 3-β-glucopyranoside, Cyanidin 3-β-glucoside, Cyanidol 3-glucoside, Glucocyanidin, Kuromanin, Kuromanine
IUPAC-namn (2S,3R,4S,5S,6R)-2-[2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromenylium-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;chloride
LEDER [Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O)cc(O)cc3[o+]c2c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material Cyanidin-3-O-glucoside chloride (Kuromanin chloride)
Kvantitet 5 mg
InChI formel InChI=1S/C21H20O11.ClH/c22-7-16-17(27)18(28)19(29)21(32-16)31-15-6-10-12(25)4-9(23)5-14(10)30-20(15)8-1-2-11(24)13(26)3-8;/h1-6,16-19,21-22,27-29H,7H2,(H3-,23,24,25,26);1H/t16-,17-,18+,19-,21-;/m1./s1
Formel vikt 484.0772