Läs mer
Cyanine 3 Monofunctional MTSEA Dye, Potassium Salt, TRC
Produktkod. 30350383
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30350383 5 mg
1 options
Produktkod. 30350383 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9605005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cyanine 3 Monofunctional MTSEA Dye, Potassium Salt
CAS 1381807-52-8
Molekylformel C34H44KN3O9S4
Kvantitet 5 mg
Synonym Cy 3 MTSEA Potassium Salt, 2-[3-[1,3-Dihydro-3,3-dimethyl-1-[6-[[2-[(methylsulfonyl)thio]ethyl]amino]-6-oxohexyl]-5-sulfo-2H-indol-2-ylidene]-1-propen-1-yl]-1-ethyl-3,3-dimethyl-5-sulfo-3H-indolium Inner Salt Potassium Salt
InChI formel InChI=1S/C34H45N3O9S4.K/c1-7-36-28-17-15-24(49(41,42)43)22-26(28)33(2,3)30(36)12-11-13-31-34(4,5)27-23-25(50(44,45)46)16-18-29(27)37(31)20-10-8-9-14-32(38)35-19-21-47-48(6,39)40;/h11-13,15-18,22-23H,7-10,14,19-21H2,1-6H3,(H2-,35,38,41,42,43,44,45,46);/q;+1/p-1
LEDER CC1(C)C(/C=C/C=C2C(C)(C)C3=CC(S(=O)(O[K])=O)=CC=C3N\2CC)=[N+](CCCCCC(NCCSS(C)(=O)=O)=O)C4=C1C=C(S(=O)([O-])=O)C=C4
IUPAC-namn potassium;2-[3-(1-ethyl-3,3-dimethyl-5-sulfonatoindol-1-ium-2-yl)prop-2-enylidene]-3,3-dimethyl-1-[6-(2-methylsulfonylsulfanylethylamino)-6-oxohexyl]indole-5-sulfonate
Molekylvikt (g/mol) 806.09
Formel vikt 805.1597