Läs mer
Cyano(1,3-dihydro-2H-benzimidazol-2-ylidene)-acetic Acid Ethyl Ester, TRC
Produktkod. 30352788
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30352788 5 mg
1 options
Produktkod. 30352788 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9791055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cyano(1,3-dihydro-2H-benzimidazol-2-ylidene)-acetic Acid Ethyl Ester
CAS 59591-86-5
Molekylformel C12H11N3O2
Kvantitet 5 mg
Synonym 2-Cyano-2-(1,3-dihydro-2H-benzimidazol-2-ylidene)-acetic Acid Ethyl Ester
LEDER NNc1ccc(Cc2ccc(NN)cc2)cc1
Molekylvikt (g/mol) 229.23