Läs mer
Cyanoguanidine-15N4, TRC
Produktkod. 30010447
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010447 5 mg
30011243 1 mg
30011250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010447 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9813525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 63419-77-2
Molekylformel C2 H4 15N4
Molekylvikt (g/mol) 88.0536
Synonym N-Cyanoguanidine-15N4, 1-Cyanoguanidine-15N4, 2-Cyanoguanidine-15N4, CGNA-15N4, Dicyandiamide-15N4, Dicyanodiamide-15N4, Didin-15N4, Epicure DICY-15N4
IUPAC-namn 1-((^{15}N)azanylidynemethyl)guanidine
LEDER [15NH2]C(=[15NH])[15NH]C#[15N]
Specifikationer
Kemiskt namn eller material Cyanoguanidine-15N4
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C2H4N4/c3-1-6-2(4)5/h(H4,4,5,6)/i3+1,4+1,5+1,6+1
Formel vikt 88.0317
Procent renhet >95%