Läs mer
Cyanomethyl Methyl(phenyl)carbamodithioate, TRC
Produktkod. 30363430
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30363430 1 g
1 options
Produktkod. 30363430 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9992381G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cyanomethyl Methyl(phenyl)carbamodithioate
CAS 76926-16-4
Molekylformel C10H10N2S2
Kvantitet 1 g
InChI formel InChI=1S/C10H10N2S2/c1-12(10(13)14-8-7-11)9-5-3-2-4-6-9/h2-6H,8H2,1H3
LEDER CN(C(=S)SCC#N)c1ccccc1
IUPAC-namn cyanomethyl N-methyl-N-phenylcarbamodithioate
Molekylvikt (g/mol) 222.32
Formel vikt 222.0285