Läs mer
Tocris Bioscience™ Cyanopindolol hemifumarate
GREENER_CHOICE
Produktkod. 18053792
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18053792 50 mg
18025272 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18053792 Leverantör Tocris Bioscience™ Leverantörsnummer 0993/50
5-HT1A and 5-HT1B antagonist; also β-adrenergic antagonist
5-HT1A/1B antagonist with roughly equal affinity at each receptor; also a β-adrenoceptor antagonist.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 874882-72-1
Molekylformel C20H25N3O5
Specifikationer
Kemiskt namn eller material Cyanopindolol hemifumarate
Kvantitet 50 mg
Formel vikt 345.4
Renhet >98%
Molekylvikt (g/mol) 387.436