Läs mer
Cyanuric Acid-13C3, TRC
Produktkod. 30009533
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009533 5 mg
30005371 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009533 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9877175MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 201996-37-4
Molekylformel 13C3 H3 N3 O3
Molekylvikt (g/mol) 132.05
Synonym 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione-2,4,6-13C3 (9CI), Cyanuric Acid 13C3
IUPAC-namn (2,4,6-^{13}C_{3})1,3,5-triazinane-2,4,6-trione
LEDER O=[13C]1N[13C](=O)N[13C](=O)N1
Specifikationer
Kemiskt namn eller material Cyanuric Acid-13C3
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C3H3N3O3/c7-1-4-2(8)6-3(9)5-1/h(H3,4,5,6,7,8,9)/i1+1,2+1,3+1
Formel vikt 132.0275
Procent renhet >95%