Läs mer
Cyclacillin, TRC
Produktkod. 30349675
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30349675 1 g
1 options
Produktkod. 30349675 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9897301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ciclacillin
CAS 3485-14-1
Molekylformel C15 H23 N3 O4 S
Kvantitet 1 g
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(1-aminocyclohexyl)carbonyl]amino]-3,3-dimethyl-7-oxo-, (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-(1-aminocyclohexanecarboxamido)-3,3-dimethyl-7-oxo- (7CI,8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(1-aminocyclohexyl)carbonyl]amino]-3,3-dimethyl-7-oxo-, [2S-(2α,5α,6β)]-, (1-Aminocyclohexyl)penicillin, 6-(1-Aminocyclohexanecarboxamido)penicillanic acid, 6-(1-Aminocyclohexylcarboxamido)penicillanic acid, Aminocyclohexylpenicillin, Calthor, Ciclacillin, Ciclacillum, Citosarin, Cyclacillin, Cyclapen, NSC 88789, Penicillin, (aminocyclohexyl)-, Syngacillin, Ultracillin, Vastcillin, Vatracin, Vipicil, Wy 4508, Wyvital
InChI formel InChI=1S/C15H23N3O4S/c1-14(2)9(12(20)21)18-10(19)8(11(18)23-14)17-13(22)15(16)6-4-3-5-7-15/h8-9,11H,3-7,16H2,1-2H3,(H,17,22)(H,20,21)/t8-,9+,11-/m1/s1
LEDER CC1(C)S[C@@H]2[C@H](NC(=O)C3(N)CCCCC3)C(=O)N2[C@H]1C(=O)O
IUPAC-namn (2S,5R,6R)-6-[(1-aminocyclohexanecarbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molekylvikt (g/mol) 341.43
Formel vikt 341.1409