Läs mer
Cyclanilide, TRC
Produktkod. 30351901
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351901 250 mg
30367038 500 mg
30366395 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30351901 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC987765250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 113136-77-9
Molekylformel C11 H9 Cl2 N O3
Molekylvikt (g/mol) 274.1
Synonym 1-[[(2,4-Dichlorophenyl)amino]carbonyl]cyclopropanecarboxylic acid, Cyclanilide, RPA 90946
IUPAC-namn 1-[(2,4-dichlorophenyl)carbamoyl]cyclopropane-1-carboxylic acid
LEDER OC(=O)C1(CC1)C(=O)Nc2ccc(Cl)cc2Cl
Specifikationer
Kemiskt namn eller material Cyclanilide
Kvantitet 250 mg
InChI formel InChI=1S/C11H9Cl2NO3/c12-6-1-2-8(7(13)5-6)14-9(15)11(3-4-11)10(16)17/h1-2,5H,3-4H2,(H,14,15)(H,16,17)
Formel vikt 272.9959