Läs mer
(-)-Cyclazocine, TRC
Produktkod. 30361093
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30361093 5 mg
1 options
Produktkod. 30361093 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9877405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-Cyclazocine
CAS 7313-86-2
Molekylformel C18 H25 N O
Kvantitet 5 mg
Synonym (2R,6R,11R)-3-(Cyclopropylmethyl)-1,2,3,4,5,6-hexahydro-6,11-dimethyl-2,6-methano-3-benzazocin-8-ol, (-)-cis-Cyclazocine, (-)-α-Cyclazocine, MCV 4512, NIH 10450, l-Cyclazocine
InChI formel InChI=1S/C18H25NO/c1-12-17-9-14-5-6-15(20)10-16(14)18(12,2)7-8-19(17)11-13-3-4-13/h5-6,10,12-13,17,20H,3-4,7-9,11H2,1-2H3/t12-,17-,18-/m1/s1
LEDER C[C@@H]1[C@H]2Cc3ccc(O)cc3[C@]1(C)CCN2CC4CC4
IUPAC-namn (1R,9S)-10-(cyclopropylmethyl)-1,13-dimethyl-10-azatricyclo[7.3.1.02,7]trideca-2(7),3,5-trien-4-ol
Molekylvikt (g/mol) 271.397
Formel vikt 271.194