missing translation for 'onlineSavingsMsg'
Läs mer

Cyclic 17-(1,2-Ethanediyl Acetal) Androst-5-ene-3,17-dione, TRC

Product Code. 30351672
Change view
Click to view available options
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30351672 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351672 Supplier Toronto Research Chemicals Supplier No. TRCC990005250MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Cyclic 17-(1,2-Ethanediyl Acetal) Androst-5-ene-3,17-dione
CAS 14456-13-4
Molekylformel C21H30O3
Kvantitet 250 mg
InChI formel InChI=1S/C21H30O3/c1-19-8-5-15(22)13-14(19)3-4-16-17(19)6-9-20(2)18(16)7-10-21(20)23-11-12-24-21/h3,16-18H,4-13H2,1-2H3/t16-,17+,18+,19+,20+/m1/s1
LEDER O=C1CC2=CC[C@@]3([H])[C@@]4([H])[C@@](C5(OCCO5)CC4)(CC[C@]3([H])[C@]2(CC1)C)C
IUPAC-namn (8R,9S,10R,13S,14S)-10,13-dimethyl-1,4,7,8,9,10,11,12,13,14,15,16-dodecahydrospiro[cyclopenta[a]phenanthrene-17,2'-[1,3]dioxolan]-3(2H)-one
Molekylvikt (g/mol) 330.46
Formel vikt 330.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.