missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10718-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with CDK7 kinase and ring finger protein MAT1. The kinase complex is able to phosphorylate CDK2 and CDC2 kinases, thus functions as a CDK-activating kinase (CAK). This cyclin and its kinase partner are components of TFIIH, as well as RNA polymerase II protein complexes. They participate in two different transcriptional regulation processes, suggesting an important link between basal transcription control and the cell cycle machinery.Specifications
Cyclin H | |
Polyclonal | |
Unconjugated | |
CCNH | |
CAK, CCNH, cyclin H, MO15 associated protein, p34, p37 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66671, 84389, 902 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51946, Q61458, Q9R1A0 | |
CCNH | |
Cyclin H Fusion Protein Ag1088 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |